2-Ethoxy-3-methylpyridine-5-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (6-Ethoxy-5-methylpyridin-3-yl)boronic acid

A887371-1G A887371-250MG A887371-5G
76820-876EA 86 USD
76820-876 76820-878 76820-880
2-Ethoxy-3-methylpyridine-5-boronic acid 98%
2-Ethoxy-3-methylpyridine-5-boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₃
MDL Number: MFCD13181608
CAS Number: 1451391-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR