5-Carboxy-2-(hydroxymethyl)benzeneboronic acid dehydrate 97%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dihydro-1-hydroxy-2,1-benzoxaborole-6-carboxylic acid, 1-Hydroxy-1,3-dihydrobenzo[c][1,2]oxaborole-6-carboxylic acid, 6-Carboxy-1,3-dihydro-1-hydroxy-2,1-benzoxaborole

A885286-100MG A885286-1G A885286-250MG
76820-866EA 73.21 USD
76820-866 76820-868 76820-870
5-Carboxy-2-(hydroxymethyl)benzeneboronic acid dehydrate 97%
5-Carboxy-2-(hydroxymethyl)benzeneboronic acid dehydrate

Precaución: for Research Use Only

Formula: C₈H₇BO₄
MW: 177.95 g/mol
MDL Number: MFCD20527893
CAS Number: 1221343-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR