Sulfacetamide sodium monohydrate 99%

Supplier: Ambeed
Warning

A885049-100MG A885049-25G A885049-5G
76820-864EA 6 USD
76820-864 77674-316 77884-830
Sulfacetamide sodium monohydrate 99%
Sulfacetamide sodium monohydrate

Precaución: for Research Use Only

Formula: C₈H₉N₂NaO₃S·H₂O
MDL Number: MFCD00149555
CAS Number: 6209-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR