Cyclopropanecarboxylic acid anhydride 90%

Supplier: Ambeed
Warning

Synonyms: Cyclopropanecarboxylic anhydride

A884149-1G A884149-250MG A884149-25G A884149-5G
76820-856EA 36.08 USD
76820-856 76820-858 76820-860 76820-862
Cyclopropanecarboxylic acid anhydride 90%
Cyclopropanecarboxylic acid anhydride

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MW: 154.163 g/mol
MDL Number: MFCD12404941
CAS Number: 33993-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR