4-Bromopyridine-2-sulfonyl chloride 98%

Supplier: Ambeed
Danger

A878208-250MG A878208-1G
76820-848EA 203.7 USD
76820-848 77573-772
4-Bromopyridine-2-sulfonyl chloride 98%
4-Bromopyridine-2-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₅H₃BrClNO₂S
MW: 256.50 g/mol
MDL Number: MFCD11847757
CAS Number: 1060808-87-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR