4,4-Difluoroazepane hydrochloride 98%

Supplier: Ambeed
Warning

A877030-100MG A877030-250MG A877030-1G
76820-846EA 41 USD
76820-846 77614-574 77884-821
4,4-Difluoroazepane hydrochloride 98%
4,4-Difluoroazepane hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁F₂N·ClH
MDL Number: MFCD11870210
CAS Number: 1160721-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR