2-Methylresorcinol 98%

Supplier: Ambeed
Danger

Synonyms: 2,6-Dihydroxytoluene, 2-Methylbenzene-1,3-diol

A876608-10G A876608-25G A876608-100G
76820-842EA 6 USD
76820-842 76820-844 77674-304
2-Methylresorcinol 98%
2-Methylresorcinol

Precaución: for Research Use Only

Formula: C₇H₈O₂
MW: 124.14 g/mol
MDL Number: MFCD00002271
CAS Number: 608-25-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR