(3-(Hydroxymethyl)-4-methylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

A875502-100MG A875502-1G A875502-250MG A875502-5G
76820-834EA 10 USD
76820-834 76820-836 76820-838 76820-840
(3-(Hydroxymethyl)-4-methylphenyl)boronic acid 98%
(3-(Hydroxymethyl)-4-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MDL Number: MFCD11617258
CAS Number: 1451391-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR