H-D-Arg(NO2)-OH 98%

Supplier: Ambeed
Danger

A875434-100G A875434-10G A875434-1G A875434-25G A875434-5G
76820-824EA 208 USD
76820-824 76820-826 76820-828 76820-830 76820-832
H-D-Arg(NO2)-OH 98%
H-D-Arg(NO2)-OH

Precaución: for Research Use Only

Formula: C₆H₁₃N₅O₄
MDL Number: MFCD00066013
CAS Number: 66036-77-9
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR