2-Hydroxy-5-pyridineboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-ol, 6-Hydroxypyridine-3-boronic acid pinacol ester

A872110-100MG A872110-1G A872110-250MG A872110-5G A872110-25G
76820-812EA 11 USD
76820-812 76820-814 76820-816 76820-818 77884-818
2-Hydroxy-5-pyridineboronic acid pinacol ester 97%
2-Hydroxy-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
MDL Number: MFCD06795671
CAS Number: 1054483-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR