2,4-Dioxo-1,3,8-triazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
8-Boc-2,4,8-triazaspiro[4.5]decane-1,3-dione, 2,4-Dioxo-1,3,8-triazaspiro[4.5]decane, N8-BOC protected
A855581-10G
A855581-1G
A855581-5G
A855581-25G
76820-760EA
139
USD
76820-760
76820-762
76820-764
77884-789
2,4-Dioxo-1,3,8-triazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester 98%
2,4-Dioxo-1,3,8-triazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₉N₃O₄ |
MDL Number:
MFCD03425517 CAS Number: 183673-70-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...