Diethyl 2,5-diaminoterephthalate 95%

Supplier: Ambeed
Warning

A852187-100MG A852187-1G A852187-250MG
76820-744EA 23.34 USD
76820-744 76820-746 76820-748
Diethyl 2,5-diaminoterephthalate 95%
Diethyl 2,5-diaminoterephthalate

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₄
MW: 252.26 g/mol
MDL Number: MFCD00629358
CAS Number: 15403-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR