2-Bromo-6-methylnicotinaldehyde 97%

Supplier: Ambeed
Danger

A847925-100MG A847925-1G A847925-250MG
76820-738EA 46.68 USD
76820-738 76820-740 76820-742
2-Bromo-6-methylnicotinaldehyde 97%
2-Bromo-6-methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆BrNO
MW: 200.03 g/mol
MDL Number: MFCD09997894
CAS Number: 853179-74-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR