(±)-2,3-Dibromopropionic acid 97%

Supplier: Ambeed
Danger

A847550-25G A847550-500G A847550-5G A847550-100G
76820-726EA 15 USD
76820-726 76820-728 76820-730 77884-780
(±)-2,3-Dibromopropionic acid 97%
(±)-2,3-Dibromopropionic acid

Precaución: for Research Use Only

Formula: BrH₂CBrCHCOOH
MDL Number: MFCD00004212
CAS Number: 600-05-5
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR