1,2-Benzisoxazole-3-methanesulfonate Sodium Salt 95%

Supplier: Ambeed
Warning

A845938-100G A845938-25G A845938-5G
76820-720EA 105.03 USD
76820-720 76820-722 76820-724
1,2-Benzisoxazole-3-methanesulfonate Sodium Salt 95%
1,2-Benzisoxazole-3-methanesulfonate Sodium Salt

Precaución: for Research Use Only

Formula: C₈H₆NNaO₄S
MW: 235.19 g/mol
MDL Number: MFCD07782146
CAS Number: 73101-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR