(R)-(1-Methylpyrrolidin-3-yl)methanol 98%

Supplier: Ambeed
Danger

A837091-5G A837091-25G
76820-694EA 165 USD
76820-694 77884-776
(R)-(1-Methylpyrrolidin-3-yl)methanol 98%
(R)-(1-Methylpyrrolidin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MDL Number: MFCD17015956
CAS Number: 1210935-33-3
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR