3,5-Dinitrosalicylic acid 97%

Supplier: Ambeed
Danger

Synonyms: 2-Hydroxy-3,5-dinitrobenzoic acid, 3,5-Dinitro-2-hydroxybenzoic acid

A834771-25G A834771-500G
76820-690EA 10.61 USD
76820-690 76820-692
3,5-Dinitrosalicylic acid 97%
3,5-Dinitrosalicylic acid

Precaución: for Research Use Only

Formula: C₇H₄N₂O₇
MW: 228.12 g/mol
MDL Number: MFCD00007104
CAS Number: 609-99-4
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR