(5-Bromopyridin-3-yl)(pyrrolidin-1-yl)methanone 98%

Supplier: Ambeed
Warning

A834292-1G A834292-25G A834292-5G
76820-684EA 17 USD
76820-684 76820-686 76820-688
(5-Bromopyridin-3-yl)(pyrrolidin-1-yl)methanone 98%
(5-Bromopyridin-3-yl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrN₂O
MDL Number: MFCD10000776
CAS Number: 1090388-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR