(4-(Hydroxymethyl)-3-methylphenyl)boronic acid 96%

Supplier: Ambeed
Warning

A833001-1G A833001-250MG A833001-5G
76820-678EA 18 USD
76820-678 76820-680 76820-682
(4-(Hydroxymethyl)-3-methylphenyl)boronic acid 96%
(4-(Hydroxymethyl)-3-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MDL Number: MFCD09996880
CAS Number: 1218790-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR