2-Chlorobenzylamine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(2-Chlorophenyl)methanamine
A830360-10G
A830360-1G
A830360-25G
A830360-5G
76820-670EA
8.49
USD
76820-670
76820-672
76820-674
76820-676
2-Chlorobenzylamine 98%
2-Chlorobenzylamine
Precaución: for Research Use Only
Formula:
C₇H₈ClN MW: 141.6 g/mol |
MDL Number:
MFCD00008108 CAS Number: 89-97-4 UN: 2735 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...