(5-Bromopicolinoyl)piperidine 98%
Supplier: Ambeed
|
Warning
|
A829814-100MG
A829814-1G
A829814-250MG
A829814-5G
76820-662EA
6
USD
76820-662
76820-664
76820-666
76820-668
(5-Bromopicolinoyl)piperidine 98%
(5-Bromopicolinoyl)piperidine
Precaución: for Research Use Only
Formula:
C₁₁H₁₃BrN₂O |
MDL Number:
MFCD08458472 CAS Number: 934000-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...