(R)-1-Boc-3-aminoazepane 98%

Supplier: Ambeed
Warning

A824902-100MG A824902-250MG A824902-1G
76820-658EA 39.26 USD
76820-658 76820-660 77674-222
(R)-1-Boc-3-aminoazepane 98%
(R)-1-Boc-3-aminoazepane

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.30 g/mol
MDL Number: MFCD11041391
CAS Number: 1032684-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR