Ethyl 2-sulfanyl-1H-imidazole-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-mercapto-1H-imidazole-4-carboxylate, 4-(Ethoxycarbonyl)-2-sulphanyl-1H-imidazole

A817150-100G A817150-1G A817150-250MG A817150-25G A817150-5G
76820-636EA 1901.14 USD
76820-636 76820-638 76820-640 76820-642 76820-644
Ethyl 2-sulfanyl-1H-imidazole-4-carboxylate 95%
Ethyl 2-sulfanyl-1H-imidazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂S
MW: 172.205 g/mol
MDL Number: MFCD00068042
CAS Number: 64038-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR