5-Chloro-2-thiophenesulfonamide 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chlorothiophene-2-sulphonamide, 2-Chloro-5-sulphamoylthiophene

A816478-100G A816478-1G A816478-25G A816478-5G
76820-628EA 237.65 USD
76820-628 76820-630 76820-632 76820-634
5-Chloro-2-thiophenesulfonamide 95%
5-Chloro-2-thiophenesulfonamide

Precaución: for Research Use Only

Formula: C₄H₄ClNO₂S₂
MW: 197.65 g/mol
MDL Number: MFCD00052584
CAS Number: 53595-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR