4-Bromo-2-fluorobenzamide 96%

Supplier: Ambeed
Warning

A816117-1G A816117-250MG A816117-25G A816117-5G
76820-620EA 6.37 USD
76820-620 76820-622 76820-624 76820-626
4-Bromo-2-fluorobenzamide 96%
4-Bromo-2-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₅BrFNO
MW: 218.03 g/mol
MDL Number: MFCD03094432
CAS Number: 292621-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR