2,2-Difluoropropane-1,3-diol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2-Difluoro-1,3-dihydroxypropane
A813054-100MG
A813054-1G
A813054-250MG
A813054-5G
76820-610EA
22.28
USD
76820-610
76820-612
76820-614
76820-616
2,2-Difluoropropane-1,3-diol 98%
2,2-Difluoropropane-1,3-diol
Precaución: for Research Use Only
Formula:
C₃H₆F₂O₂ MW: 112.075 g/mol |
MDL Number:
MFCD16619633 CAS Number: 428-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...