Imidazo[1,2-a]pyridine-5-carboxylic acid hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A812140-100MG
A812140-1G
A812140-250MG
76820-604EA
31
USD
76820-604
76820-606
76820-608
Imidazo[1,2-a]pyridine-5-carboxylic acid hydrochloride 98%
Imidazo[1,2-a]pyridine-5-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₇ClN₂O₂ |
MDL Number:
MFCD09954931 CAS Number: 1198283-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...