5-(Hydroxymethyl)oxazolidin-2-one 97%
Supplier: Ambeed
|
Warning
|
A810218-1G
A810218-250MG
A810218-5G
76820-598EA
88.06
USD
76820-598
76820-600
76820-602
5-(Hydroxymethyl)oxazolidin-2-one 97%
5-(Hydroxymethyl)oxazolidin-2-one
Precaución: for Research Use Only
Formula:
C₄H₇NO₃ MW: 117.10 g/mol |
MDL Number:
MFCD05721226 CAS Number: 7517-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...