(4-Bromophenyl)diphenylphosphine 98%

Supplier: Ambeed
Warning

A809917-250MG A809917-1G
76820-596EA 44.56 USD
76820-596 77614-442
(4-Bromophenyl)diphenylphosphine 98%
(4-Bromophenyl)diphenylphosphine

Precaución: for Research Use Only

Formula: C₁₈H₁₄BrP
MW: 341.18 g/mol
MDL Number: MFCD00000099
CAS Number: 734-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR