4,4'-Diiodo-2,2'-dimethylbiphenyl 95%

Supplier: Ambeed
Warning

A808261-10G A808261-1G A808261-250MG A808261-5G
76820-582EA 82 USD
76820-582 76820-584 76820-586 76820-588
4,4'-Diiodo-2,2'-dimethylbiphenyl 95%
4,4'-Diiodo-2,2'-dimethylbiphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂I₂
MDL Number: MFCD13185944
CAS Number: 69571-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR