Methyl (4-nitrophenyl)acetate 98%

Supplier: Ambeed
Warning

A807444-100G A807444-100MG A807444-1G A807444-5G A807444-10G A807444-25G
76820-574EA 342 USD
76820-574 76820-576 76820-578 76820-580 77884-753 77884-754
Methyl (4-nitrophenyl)acetate 98%
Methyl (4-nitrophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD00024802
CAS Number: 2945-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR