4-(Hydroxymethyl)cyclohexanol (cis- and trans- mixture) 98%

Supplier: Ambeed
Warning

A805687-100MG A805687-1G A805687-5G
76820-572EA 18 USD
76820-572 77674-186 77884-750
4-(Hydroxymethyl)cyclohexanol (cis- and trans- mixture) 98%

Precaución: for Research Use Only

Formula: C₇H₁₄O₂
MDL Number: MFCD11036248
CAS Number: 33893-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR