Pyrrolidine-3-carboxamide hydrochloride 98%

Supplier: Ambeed
Warning

A802792-100MG A802792-1G A802792-250MG
76820-556EA 58.35 USD
76820-556 76820-558 76820-560
Pyrrolidine-3-carboxamide hydrochloride 98%
Pyrrolidine-3-carboxamide hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁ClN₂O
MW: 150.60 g/mol
MDL Number: MFCD17977027
CAS Number: 644972-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR