3-Chloro-2-isobutoxy-5-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (5-Chloro-6-isobutoxypyridin-3-yl)boronic acid

A802362-100MG A802362-1G A802362-250MG
76820-546EA 36.08 USD
76820-546 76820-548 76820-550
3-Chloro-2-isobutoxy-5-pyridineboronic acid 98%
3-Chloro-2-isobutoxy-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BClNO₃
MW: 229.4700 g/mol
MDL Number: MFCD12546521
CAS Number: 1217500-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR