4-(Benzyloxy)-3-fluorobenzaldehyde 95%

Supplier: Ambeed
Warning

A800249-250MG A800249-5G
76820-542EA 36 USD
76820-542 76820-544
4-(Benzyloxy)-3-fluorobenzaldehyde 95%
4-(Benzyloxy)-3-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO₂
MDL Number: MFCD11156437
CAS Number: 175968-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR