1-(4-Methoxyphenyl)cyclobutanecarbonitrile 98+%

Supplier: Ambeed
Warning

A792574-100MG A792574-1G A792574-250MG
76820-510EA 13.8 USD
76820-510 76820-512 76820-514
1-(4-Methoxyphenyl)cyclobutanecarbonitrile 98+%
1-(4-Methoxyphenyl)cyclobutanecarbonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO
MDL Number: MFCD00297175
CAS Number: 29786-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR