4-Bromo-5-ethyl-2-methylpyridine 95%

Supplier: Ambeed
Danger

A788809-1G A788809-250MG
76820-500EA 128 USD
76820-500 76820-502
4-Bromo-5-ethyl-2-methylpyridine 95%
4-Bromo-5-ethyl-2-methylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.07 g/mol
MDL Number: MFCD21603856
CAS Number: 98488-99-4
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR