3-Bromo-5-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A788547-10G A788547-1G A788547-250MG
76820-494EA 403.15 USD
76820-494 76820-496 76820-498
3-Bromo-5-nitrobenzonitrile 98%
3-Bromo-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrN₂O₂
MDL Number: MFCD00463804
CAS Number: 49674-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR