N-Benzoyl-L-glutamic acid 98%

Supplier: Ambeed
Warning

Synonyms: N-Bz-Glu-OH, Benzoyl-L-glutamic acid, Bz-Glu-OH

A784189-100G A784189-5G
76820-470EA 191 USD
76820-470 76820-472
N-Benzoyl-L-glutamic acid 98%
N-Benzoyl-L-glutamic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₅
MW: 251.24 g/mol
MDL Number: MFCD00014028
CAS Number: 6094-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR