3-Amino-5,6-dichloropyrazine-2-carboxamide 95+%

Supplier: Ambeed
Danger

A782848-100MG A782848-1G A782848-250MG
76820-460EA 41.38 USD
76820-460 76820-462 76820-464
3-Amino-5,6-dichloropyrazine-2-carboxamide 95+%
3-Amino-5,6-dichloropyrazine-2-carboxamide

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₄O
MW: 207.01 g/mol
MDL Number: MFCD21603848
CAS Number: 14229-27-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR