2,3-Difluoro-4-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Danger

A780696-250MG A780696-1G A780696-5G
76820-458EA 17 USD
76820-458 77884-718 77884-719
2,3-Difluoro-4-(trifluoromethyl)pyridine 97%
2,3-Difluoro-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂F₅N
MDL Number: MFCD16610846
CAS Number: 1215034-77-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR