N-(2-Acetamido)-2-aminoethanesulfonic acid (ACES) 99%

Supplier: Ambeed
Warning

Synonyms: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid

A780410-100G A780410-25G A780410-500G
76820-452EA 30.77 USD
76820-452 76820-454 76820-456
N-(2-Acetamido)-2-aminoethanesulfonic acid (ACES) 99%
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES)

Precaución: for Research Use Only

Formula: H₂NCOCH₂NHCH₂CH₂SO₃H
MW: 182.2 g/mol
MDL Number: MFCD00008030
CAS Number: 7365-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR