3-Methoxy-5-(trifluoromethoxy)phenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (3-Methoxy-5-(trifluoromethoxy)phenyl)boronic acid

A775004-100MG A775004-1G A775004-250MG A775004-5G
76820-442EA 17 USD
76820-442 76820-444 76820-446 76820-448
3-Methoxy-5-(trifluoromethoxy)phenylboronic acid 96%
3-Methoxy-5-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₄
MW: 235.9529 g/mol
MDL Number: MFCD12026733
CAS Number: 1256345-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR