DL-ɑ-Amino-ε-caprolactam 98%

Supplier: Ambeed
Warning

Synonyms: DL-3-Amino-2-oxohexamethyleneimine, DL-3-Aminohexahydro-2-azepinone, 3-Amino-2-azepanone(Chunks or pellets or lump), 3-Amino-2-azepanone

A773925-100G A773925-1G A773925-25G A773925-5G
76820-434EA 818 USD
76820-434 76820-436 76820-438 76820-440
DL-ɑ-Amino-ε-caprolactam 98%
DL-alpha-Amino epsilon caprolactam

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O
MW: 128.18 g/mol
MDL Number: MFCD00064475
CAS Number: 671-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR