N,N-Dimethyl 3-boronobenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: (3-(N,N-Dimethylsulphamoyl)phenyl)boronic acid, 3-(Dimethylsulphamoyl)benzeneboronic acid

A773479-100MG A773479-1G A773479-250MG
76820-424EA 37.14 USD
76820-424 76820-426 76820-428
N,N-Dimethyl 3-boronobenzenesulfonamide 98%
N,N-Dimethyl 3-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₄S
MW: 229.06 g/mol
MDL Number: MFCD07363747
CAS Number: 871329-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR