5-Bromo-4-carboxy-2-fluorobenzeneboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-carboxy-2-fluorophenylboronic acid, 4-Borono-2-bromo-5-fluorobenzoic acid

A771083-1G A771083-250MG A771083-5G
76820-404EA 129 USD
76820-404 76820-406 76820-408
5-Bromo-4-carboxy-2-fluorobenzeneboronic acid 96%
5-Bromo-4-carboxy-2-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BBrFO₄
MDL Number: MFCD09475847
CAS Number: 957034-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR