1-tert-Butyl-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butyl)-1H-pyrazole

A767253-10G A767253-1G A767253-5G
76820-386EA 147 USD
76820-386 76820-388 77674-108
1-tert-Butyl-1H-pyrazole 97%
1-tert-Butyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₁₂N₂
MDL Number: MFCD12192641
CAS Number: 15754-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR