2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]oxazole 97%

Supplier: Ambeed
Danger

A766178-100MG A766178-1G A766178-250MG A766178-5G
76820-378EA 42.44 USD
76820-378 76820-380 76820-382 76820-384
2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]oxazole 97%
2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]oxazole

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₃
MW: 259.10 g/mol
MDL Number: MFCD13181970
CAS Number: 845872-30-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR