5-Bromopyridazin-3-amine 98%

Supplier: Ambeed
Warning

A764211-100MG A764211-1G
76820-376EA 238 USD
76820-376 77552-348
5-Bromopyridazin-3-amine 98%
5-Bromopyridazin-3-amine

Precaución: for Research Use Only

Formula: C₄H₄BrN₃
MW: 173.99 g/mol
MDL Number: MFCD18382564
CAS Number: 1187237-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR