3-(2,2,2-Trifluoroethoxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(2,2,2-Trifluoroethoxy)benzeneboronic acid, (3-(2,2,2-Trifluoroethoxy)phenyl)boronic acid

A762263-100MG A762263-250MG A762263-5G
76820-354EA 38 USD
76820-354 76820-356 76820-358
3-(2,2,2-Trifluoroethoxy)phenylboronic acid 98%
3-(2,2,2-Trifluoroethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₃
MDL Number: MFCD07363846
CAS Number: 850593-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR